Order Entry
Export
Orders LinkContactUsLinkComponent
Human CD63 ELISA Kit

Human CD63 ELISA Kit

Human CD63 ELISA Kit, 96 tests
  ABCAAB275099-1X96
 :   Abcam
Human CD63 ELISA Kit
  ABCAAB275099-1X96
 :  Abcam
 :  
Human CD63 ELISA Kit
Human CD63 ELISA Kit

Human CD63 ELISA Kit, 96 tests

 :  
 
723,00 €
Log in for availability
1 * 1 KIT
 

Human CD63 ELISA kit is a single-wash 90-min Simplestep used to quantify Human CD63 with a sensitivity of 33 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.

Specifications

Specifications

  • Description:
    Human CD63 ELISA Kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Cell culture supernatant, Serum, EDTA Plasma
  • Detection range:
    46,9 - 3000 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 hr 30 min
  • Cat. No.:
    ABCAAB275099-1X96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    33 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    CD63
  • Regulatory status:
    RUO

Specifications