Order Entry
Export
Orders LinkContactUsLinkComponent
TW-37 ≥98%

TW-37 ≥98%

Catalog # ABCAAB273490-100
Supplier:   Abcam
TW-37 ≥98%
Catalog # ABCAAB273490-100
Supplier:  Abcam
CAS Number:  877877-35-5
TW-37 ≥98%
CAS Number:  877877-35-5
Price
1 120,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Small-molecule inhibitor of BCL-2 (Ki = 0,29 μM). Inhibits multiple BCL-2 family members including BCL-XL (Ki = 1,11 μM) and MCL-1 (Ki= 0,26 μM). Binds to the BCL-2 homology domain 3 (BH3) groove of BCL-2. Significant anti-proliferative and pro-apoptotic activity in lymphoma cells.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  N-[4-(2-tert-Butylphenylsulphonyl)phenyl]-2,3,4-trihydroxy-5-(2-isopropylbenzyl)benzamide
  • CAS Number:877877-35-5
  • Molecular Weight:573,7 g/mol
Specifications

Specification Test Results

  • Constituents::
    98% TW-37