0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant Human EBI3 protein 1 * 20 µG
ABCAAB269153-20
:
Abcam
Recombinant Human EBI3 protein 1 * 20 µG
ABCAAB269153-20
:
Abcam