0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant Mouse IL-31 protein 1 * 100 µG
ABCAAB256058-100
:
Abcam
Recombinant Mouse IL-31 protein 1 * 100 µG
ABCAAB256058-100
:
Abcam