Order Entry
Export
Orders LinkContactUsLinkComponent
Rat Recombinant IL-10 (from E. coli)

Rat Recombinant IL-10 (from E. coli)

Catalog # ABCAAB256036-1
Supplier:   Abcam
Rat Recombinant IL-10 (from E. coli)
Catalog # ABCAAB256036-1
Supplier:  Abcam
CAS Number:  
Rat Recombinant IL-10 (from E. coli)
Rat Recombinant IL-10 (from E. coli)
CAS Number:  
Price
6 780,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Recombinant Rat IL-10 protein (Animal Free) is a Rat Full Length protein, in the 19 to 178 aa range, expressed in Escherichia coli, with >95% purity, <1 EU/µg endotoxin level and suitable for SDS-PAGE.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Lyophilised
  • Protein/peptide name:
    IL-10
  • Protein synonyms:
    Il10|Interleukin-10|Cytokine synthesis inhibitory factor|CSIF
  • Purity:
    >95%
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    Ambient - Can ship with ice
  • Cat. No.:
    ABCAAB256036-1
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <1 EU/µg
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    Constituents: 0,1% Trifluoroacetic acid
  • Conjugation:
    Unconjugated
  • Tested applications:
    SDS-PAGE
  • Species:
    Rat
  • Animal-free:
    Y

Specifications