Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse Interferon alpha 1 ELISA Kit

Mouse Interferon alpha 1 ELISA Kit

Mouse Interferon alpha 1 ELISA kit, 96 tests
Catalog # ABCAAB252352-1X96
Supplier:   Abcam
Mouse Interferon alpha 1 ELISA Kit
Catalog # ABCAAB252352-1X96
Supplier:  Abcam
CAS Number:  
Mouse Interferon alpha 1 ELISA Kit
Mouse Interferon alpha 1 ELISA Kit

Mouse Interferon alpha 1 ELISA kit, 96 tests

CAS Number:  
Price
810,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Mouse Interferon alpha 1 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse Interferon alpha 1 with a sensitivity of 7 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations.

Specifications

Specifications

  • Description:
    Mouse Interferon alpha 1 ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Cell culture media, Heparin Plasma, Citrate plasma, Serum
  • Detection range:
    15,6 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB252352-1X96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    7 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Interferon alpha 1
  • Regulatory status:
    RUO

Specifications