Tissue Clearing Permeabilization Buffer - hydrophobic, Abcam
Product Details & Documents
Tissue Clearing Permeabilization Buffer ab243307 is part of our range of tissue clearing reagents and products that enable you to set up tissue clearing easily and quickly.
- Constituents: 94% Water, 6% Sodium lauryl sulfate
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Description:Tissue Clearing Permeabilization Buffer - hydrophobic
- Cat. No.:ABCAAB243307-30
- Storage temperature:Store at Ambient.



