Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant FGF5 (from E. coli)

Human Recombinant FGF5 (from E. coli)

Catalog # ABCAAB219125-100
Supplier:   Abcam
Human Recombinant FGF5 (from E. coli)
Catalog # ABCAAB219125-100
Supplier:  Abcam
CAS Number:  
Human Recombinant FGF5 (from E. coli)
Human Recombinant FGF5 (from E. coli)
CAS Number:  
Price
400,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Recombinant human FGF5 protein (Active) is a Human Full Length protein, in the 18 to 268 aa range, expressed in Escherichia coli, with ≥95% purity, ≤1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Lyophilised
  • Protein/peptide name:
    FGF5
  • Protein synonyms:
    Fibroblast growth factor 5|FGF-5|Heparin-binding growth factor 5|Smag-82|HBGF-5
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Biological activity:
    Bioactive
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB219125-100
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <1 EU/µg
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    Constituents: 0,58% Sodium chloride, 0,16% Sodium phosphate
  • Conjugation:
    Unconjugated
  • Tested applications:
    FuncS, SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications