Order Entry
Export
Orders LinkContactUsLinkComponent
Mounting Medium With DAPI - Aqueous, Fluoroshield, Abcam

Mounting Medium With DAPI - Aqueous, Fluoroshield, Abcam

Mounting Medium With DAPI - Aqueous, Fluoroshield
Catalog # ABCAAB104139-100
Supplier:   Abcam
Mounting Medium With DAPI - Aqueous, Fluoroshield, Abcam
Catalog # ABCAAB104139-100
Supplier:  Abcam
CAS Number:  
Mounting Medium With DAPI - Aqueous, Fluoroshield, Abcam

Mounting Medium With DAPI - Aqueous, Fluoroshield

CAS Number:  
Price
436,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

DAPI mounting medium (previously called Fluoroshield Mounting Medium With DAPI) is an aqueous anti-fade mounting medium designed to preserve fluorescence when imaging tissues and cell samples. - Cited in over 325 publications - Suitable for immunohistochemistry (IHC), immunocytochemistry/immunofluorescence (ICC/IF) and more - Contains 84% water, 0.0002% 4',6-Diamidino-2-phenylindole dihydrochloride

  • Preservative: 0.09% Sodium azide Constituents: 84% Water, 0.0002% 4',6-Diamidino-2-phenylindole dihydrochloride

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Mounting Medium With DAPI - Aqueous, Fluoroshield
  • Cat. No.:
    ABCAAB104139-100
  • Storage temperature:
    Store at +4°C.

Specifications