0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant Human ALDH5A1SSADH protein 1 * 250 µG
ABCAAB99429-250
:
Abcam
Recombinant Human ALDH5A1SSADH protein 1 * 250 µG
ABCAAB99429-250
:
Abcam