0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant human MEK1 protein 1 * 5 µG
ABCAAB63209-5
:
Abcam
Recombinant human MEK1 protein 1 * 5 µG
ABCAAB63209-5
:
Abcam