Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse IL-2 ELISPOT Set

Mouse IL-2 ELISPOT Set

Mouse IL-2 ELISPOT set, 15×96 tests
Catalog # ABCAAB46623-15X96
Supplier:   Abcam
Mouse IL-2 ELISPOT Set
Catalog # ABCAAB46623-15X96
Supplier:  Abcam
CAS Number:  
Mouse IL-2 ELISPOT Set
Mouse IL-2 ELISPOT Set

Mouse IL-2 ELISPOT set, 15×96 tests

CAS Number:  
Price
2 350,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Mouse IL-2 ELISPOT Set is a ELISPOT Set for the sensitive detection of Mouse IL-2 production in a single cell suspension.

Specifications

Specifications

  • Description:
    Mouse IL-2 ELISPOT set
  • Tests per kit:
    15×96 tests
  • Cat. No.:
    ABCAAB46623-15X96
  • Sample type:
    Suspension cells
  • Species reactivity:
    Mouse
  • Product type:
    Uncoated
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen symbol:
    IL-2

Specifications