Order Entry
Export
Orders LinkContactUsLinkComponent
 

Mouse IL-2 ELISPOT Set

Mouse IL-2 ELISPOT set, 10×96 tests
  ABCAAB46623-10X96
 :   Abcam
Mouse IL-2 ELISPOT Set
  ABCAAB46623-10X96
 :  Abcam
 :  
Mouse IL-2 ELISPOT Set
Mouse IL-2 ELISPOT Set

Mouse IL-2 ELISPOT set, 10×96 tests

 :  
 
1 700,00 €
   
1 * 1 KIT
 

Mouse IL-2 ELISPOT Set is a ELISPOT Set for the sensitive detection of Mouse IL-2 production in a single cell suspension.

 

 

  • Description:
    Mouse IL-2 ELISPOT set
  • Tests per kit:
    10×96 tests
  • Cat. No.:
    ABCAAB46623-10X96
  • Sample type:
    Suspension cells
  • Species reactivity:
    Mouse
  • Product type:
    Uncoated
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen symbol:
    IL-2