Mouse IL-2 ELISPOT Set
Mouse IL-2 ELISPOT set, 20×96 tests
Catalog # ABCAAB46623-20X96
Supplier: Abcam
Jump to:
Product Details & Documents
Mouse IL-2 ELISPOT Set is a ELISPOT Set for the sensitive detection of Mouse IL-2 production in a single cell suspension.
Specifications
Specifications
- Description:Mouse IL-2 ELISPOT set
- Tests per kit:20×96 tests
- Cat. No.:ABCAAB46623-20X96
- Sample type:Suspension cells
- Species reactivity:Mouse
- Product type:Uncoated
- Storage temperature:4 °C
- Processing time:Multiple steps
- Regulatory status:RUO
- Antigen symbol:IL-2



