Human Granzyme B ELISPOT Set
Human Granzyme B ELISPOT set
Catalog # ABCAAB46581-20X96
Supplier: Abcam
Jump to:
Product Details & Documents
Human Granzyme B ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Granzyme B production in a single cell suspension.
Specifications
Specifications
- Description:Human Granzyme B ELISPOT set
- Cat. No.:ABCAAB46581-20X96
- Sample type:Suspension cells
- Form:Uncoated
- Species reactivity:Human
- Storage temperature:+4 °C
- Processing time:Multiple steps
- Regulatory status:RUO
- Antigen name:Granzyme B



