Order Entry
Export
Orders LinkContactUsLinkComponent
Human Interferon gamma ELISPOT Set

Human Interferon gamma ELISPOT Set

Human Interferon gamma ELISPOT set, 20×96 tests
Catalog # ABCAAB46551-20X96
Supplier:   Abcam
Human Interferon gamma ELISPOT Set
Catalog # ABCAAB46551-20X96
Supplier:  Abcam
CAS Number:  
Human Interferon gamma ELISPOT Set
Human Interferon gamma ELISPOT Set

Human Interferon gamma ELISPOT set, 20×96 tests

CAS Number:  
Price
2 790,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Human Interferon gamma ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Interferon gamma production in a single cell suspension.

Specifications

Specifications

  • Description:
    Human Interferon gamma ELISPOT set
  • Tests per kit:
    20×96 tests
  • Cat. No.:
    ABCAAB46551-20X96
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Species reactivity:
    Human
  • Storage temperature:
    4 ℃
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen name:
    Interferon gamma

Specifications