Human Interferon gamma ELISPOT Set
Human Interferon gamma ELISPOT set, 10×96 tests
ABCAAB46551-10X96
: Abcam
Human Interferon gamma ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Interferon gamma production in a single cell suspension.
- Description:Human Interferon gamma ELISPOT set
- Tests per kit:10×96 tests
- Cat. No.:ABCAAB46551-10X96
- Sample type:Suspension cells
- Form:Uncoated
- Species reactivity:Human
- Storage temperature:4 ℃
- Processing time:Multiple steps
- Regulatory status:RUO
- Antigen name:Interferon gamma



