Human Interferon gamma ELISPOT Set
Human Interferon gamma ELISPOT set, 15×96 tests
Catalog # ABCAAB46551-15X96
Supplier: Abcam
Jump to:
Product Details & Documents
Human Interferon gamma ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Interferon gamma production in a single cell suspension.
Specifications
Specifications
- Description:Human Interferon gamma ELISPOT set
- Tests per kit:15×96 tests
- Cat. No.:ABCAAB46551-15X96
- Sample type:Suspension cells
- Form:Uncoated
- Species reactivity:Human
- Storage temperature:4 ℃
- Processing time:Multiple steps
- Regulatory status:RUO
- Antigen name:Interferon gamma



