Order Entry
Export
Orders LinkContactUsLinkComponent
Human TNF alpha ELISPOT Set

Human TNF alpha ELISPOT Set

Human TNF alpha ELISPOT set, 10×96 tests
Catalog # ABCAAB46550-10X96
Supplier:   Abcam
Human TNF alpha ELISPOT Set
Catalog # ABCAAB46550-10X96
Supplier:  Abcam
CAS Number:  
Human TNF alpha ELISPOT Set
Human TNF alpha ELISPOT Set

Human TNF alpha ELISPOT set, 10×96 tests

CAS Number:  
Price
1 700,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Specifications

Specifications

  • Description:
    Human TNF alpha ELISPOT set
  • Tests per kit:
    10×96 tests
  • Cat. No.:
    ABCAAB46550-10X96
  • Form:
    Uncoated
  • Species reactivity:
    Human;Monkey
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen symbol:
    TNF alpha

Specifications