0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Recombinant Human Hsp40 protein 1 * 20 µG
ABCAAB43613-20
:
Abcam
Recombinant Human Hsp40 protein 1 * 20 µG
ABCAAB43613-20
:
Abcam