Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
50 kg
Supplier: Avantor
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Catalog # 3718-39
Supplier: Avantor
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Meets N.F. Requirements, Meets J.P. Chemical Specifications, Meets E.P. Chemical Specifications, Meets B.P. Chemical Specifications, GMP Manufactured Product
- GMP Manufactured Product
- Bulk Pharmaceutical Chemical
: For Manufacturing, processing or repackaging
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2.13 g/cm³ (20 °C)
- EINECS:215-185-5
- ADR:8,II
- :Management of Change (MOC) category = R
- Storage Temperature:Ambient
- UN:1823
- REACH:01-2119457892-27
Specifications
Specification Test Results
- NF - Identification A (Sodium):Passes Test
- NF - Identification B (pH):≥ 11.0
- NF - Insoluble Substances and Organic Matter:Passes Test
- NF - Potassium (K):≤ 0.5 %
- NF - Carbonate (as Na₂CO₃):≤ 3.0 %
- NF - Total Alkali:95.0 - 100.5 %
- NF - Content of Sodium:54.0 - 59.8 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Carbonate (as Na₂CO₃):≤ 2.0 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Sulfate (SO₄):≤ 200 ppm
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Assay (NaOH):97.0 - 100.5 %
- JP - Assay:95.0 - 101.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤ 0.050 %
- JP - Potassium (K):Passes Test
- JP - Carbonate (as Na₂CO₃):≤ 2.0 %
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:≤2.0%
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:Passes Test
Specifications
- Pk:50 kg



