Order Entry
Export
ContactUsLinkComponent
[EN]FLOW CONTROLLER 2.5L 1/2 FLARETEK 1 * 1 ST
Catalog # ENTE6500T5F4D12AP7
Supplier:  ENTEGRIS
[EN]FLOW CONTROLLER 2.5L 1/2 FLARETEK 1 * 1 ST
Catalog # ENTE6500T5F4D12AP7
Supplier:  ENTEGRIS