Orders LinkContactUsLinkComponent
Grounding cables, b.safe
Grounding cables, b.safe
Grounding cable with clip and stainless steel strap, 1,5 m
Catalog # 590-0345
Supplier:   Bohlender
Grounding cables, b.safe
Catalog # 590-0345
Supplier:  Bohlender
Grounding cables, b.safe
Grounding cables, b.safe
Grounding cable with clip and stainless steel strap, 1,5 m
Price
63,40 €
Log in for availability
1 * 1 items
Product Details & Documents
For earth connection of vessels and components made of static dissipative materials.
  • Choose the suitable connection to earth that we offer in our range of grounding cables with different connectors
  • Customised versions can be offered on request
  • Suitable grounding accessories for b.safe products
The scope of delivery of the ring cable lug includes screw, nut and washer.
Specifications
  • Description:
    Grounding cable with clip and stainless steel strap
  • Length:
    1,5 m
  • Seq:
    05