Capillary cutter, b.safe
Capillary cutter for tubing up to Ext.Ø 28 mm
590-0335
Ideal for cutting capillaries and elastic tubes without reinforcement.
- Capillary cutter for up to tubing Ext.Ø 8 mm with guide bores
- High qualitiy capillary cutter
- Replacement blades available
The guide bores help to get a clean cut of the capillaries.
- Description:Capillary cutter for tubing up to Ext.Ø 28 mm
- Seq:02



