Spouts, b.safe
Spout tube flexible, S70/71, PE
Catalog # 590-0325
Supplier: Bohlender
Jump to:
Product Details & Documents
Screw cap for canister thread, freely turnable spout tube, rigid or flexible design with integrated safety aeration for constant flow.
- Spout tube is freely turnable
- Integrated safety aeration
- Prevents loss of fluids
Quick refilling of liquids without dripping directly from canister.
Specifications
- Thread:S70/71
- Description:Spout tube flexible
- Material:PE
- Seq:04