Orders LinkContactUsLinkComponent
Spouts, b.safe
Spouts, b.safe
Spout tube flexible, S55, PE
Catalog # 590-0323
Supplier:   Bohlender
Spouts, b.safe
Catalog # 590-0323
Supplier:  Bohlender
Spouts, b.safe
Spout tube flexible, S55, PE
Price
17,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Screw cap for canister thread, freely turnable spout tube, rigid or flexible design with integrated safety aeration for constant flow.
  • Spout tube is freely turnable
  • Integrated safety aeration
  • Prevents loss of fluids
Quick refilling of liquids without dripping directly from canister.
Specifications
  • Thread:
    S55
  • Description:
    Spout tube flexible
  • Material:
    PE
  • Seq:
    02