Orders LinkContactUsLinkComponent
Waste caps, b.safe
Waste caps, b.safe
Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connectors for tubing with int. Ø 5,5 - 8,0 mm, B63
Catalog # 590-0144
Supplier:   Bohlender
Waste caps, b.safe
Catalog # 590-0144
Supplier:  Bohlender
Waste caps, b.safe
Waste caps, b.safe
Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connectors for tubing with int. Ø 5,5 - 8,0 mm, B63
Price
250,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Safe collection of hazardous waste is an important part of the laboratory work routine. b.safe waste caps connect waste lines from your HPLC/UHPLC leak-tight with your waste container. The waste caps are available in various sizes to fit your specific waste container safely.
  • Available in many thread sizes to fit a wide range of waste containers
  • Excellent chemical resistance
  • Accessories like fittings and hose connectors guarantee you waste lines are connected leak tight
  • Special couplings for corrugated flexible tubes
  • Exhaust filters for different sizes of waste containers that adsorb hazardous vapours
Suitable fittings, hose connectors and special couplings for corrugated hoses, make the b.safe waste system very flexible. Materials like PTFE, PPS, PFA and PP guarantee excellent chemical resistant. Now all you need is a b.safe exhaust filter to complete your safe waste collection system.
Safe collection of harmful solvent waste as well as perfect protection from hazardous vapours and liquids, also absolute control until non-hazardous collection of waste. This is what b.safe stands for.
After extraction and analysis have been successfully completed, lab experts cannot relax because at the end of every HPLC process, there is harmful solvent waste that increases the risk of fire and explosion and the health of employees. The waste has, therefore, to be collected in special vessels until their disposal without vapours and liquids being able to escape. A clear case for b.safe waste caps and b.safe exhaust filters.
b.safe waste caps are fixed tightly on the threads of the waste containers. Fittings, hose connectors and the practical distributor body, tubing and capillaries can be fixed without the unit being dismantled. As a result, eluents come into the waste containers safely and not outside of them. A special activated carbon in the b.safe exhaust filters absorbs all solvent vapours in order to have only harmless air coming into the environment. Practical indicators guarantee full control on permitted service life.
Specifications
  • Thread:
    B63
  • Description:
    Waste caps, 4× capillary connector, including 4× fittings for tubing with ext. Ø 1,6 mm, 4× fittings for tubing with ext. Ø 2,2 mm, 4× fittings for tubing with ext. Ø 3,2 mm, 3 blind fittings, 1 hose connectors for tubing with int. Ø 5,5 - 8,0 mm
  • Seq:
    37