Original Perfusor® syringes
Original Perfusor® Syringe 50 ml, with particle filter cannula, 50 ml, — ml, — ml, — mm, Centric cone
BRAU8728852F-06
: B. Braun
PP syringe with safe plunger backstop and minimal residual volume.
- Oval grip plate for handling of the syringe and for prevention of rolling
- Plunger stopper with double sealing ring for slow aspiration or infusion of drug solutions, made of synthetic material or natural latex-free
- Centric Luer Lock cone for the attachment of a needle or infusion tubing
- Ext.Ø:— mm
- Type:Centric cone
- Capacity:50 ml
- Description:Original Perfusor® Syringe 50 ml, with particle filter cannula
- Length:— ml
- Seq:06
- Graduation:— ml