Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Kabelbinder L: 145mm W: 3,4 mm PEEK 230N beige Max:: 35mm Arbeitstemperatur -55...240C,, 1 * 100 ST
Catalog # HELT118-00032
Supplier:
HELLERMANN TYTON
Expand image
Kabelbinder L: 145mm W: 3,4 mm PEEK 230N beige Max:: 35mm Arbeitstemperatur -55...240C,, 1 * 100 ST
Catalog # HELT118-00032
Supplier:
HELLERMANN TYTON