Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
100 ml, 14/23, 0,100 ± ml
Catalog # HIRS2820281
Supplier:   HIRSCHMANN
Volumetric flasks, class A, blue markings
Catalog # HIRS2820281
Supplier:  HIRSCHMANN
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
100 ml, 14/23, 0,100 ± ml
Price
56,40 €
Log in for availability
1 * 2 items
Product Details & Documents
DURAN®, borosilicate glass, with glass stopper, conformity certified (DE-M marking).
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification (available via website: www.hirschmannlab.de)
* In addition to DIN
**Wide neck
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    14/23
  • Capacity:
    100 ml
  • Seq:
    12
  • Tolerance:
    0,100 ± ml