Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
50 ml, 12/21, 0,060 ± ml
Catalog # HIRS2820275
Supplier:   HIRSCHMANN
Volumetric flasks, class A, blue markings
Catalog # HIRS2820275
Supplier:  HIRSCHMANN
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
50 ml, 12/21, 0,060 ± ml
Price
50,70 €
Log in for availability
1 * 2 items
Product Details & Documents
DURAN®, borosilicate glass, with glass stopper, conformity certified (DE-M marking).
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification (available via website: www.hirschmannlab.de)
* In addition to DIN
**Wide neck
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    12/21
  • Capacity:
    50 ml
  • Seq:
    09
  • Tolerance:
    0,060 ± ml