Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lithium bromide 99%

Bottle, 500 g
  APOSIN2314-500G
Irritant
Lithium bromide 99%
  APOSIN2314-500G
 :  7550-35-8
Lithium bromide 99%

Bottle, 500 g

 :  7550-35-8
 
65,20 €
Log in for availability
1 * 500 g
 
  • CAS Number:7550-35-8
  • Formula:LiBr
  • MDL Number:MFCD00011077
  • Molecular Weight:86.84 g/mol
  • Boiling Pt:1265 °C
  • Melting Pt:550 °C
  • Density:3.46 g/cm³ (25 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Cat. No.:
    APOSIN2314-500G

Specifications