(-)-Quinine
Bottle, 5 g
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
- CAS Number:130-95-0
- Formula:C₂₀H₂₄N₂O₂
- MDL Number:MFCD00198096
- Molecular Weight:324.42 g/mol
- Melting Pt:173…175 °C
- Storage Temperature:Ambient
- EINECS:205-003-2
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:5 g
- Cat. No.:APOSOR28880-5G



