: (R)-[(2S,4S,5R)-1-Aza-5-vinylbicyclo[2.2.2]oct-2-yl](6-methoxyquinolin-4-yl)methanol, Quinine, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol
- :130-95-0
- :C₂₀H₂₄N₂O₂
- :MFCD00198096
- :324.42 g/mol
- :173…175 °C
- :Ambient
- :205-003-2