Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Hexanedione 97%

2,4-Hexanedione 97%

Bottle, 1 g
Catalog # APOSOR938383-1G
Supplier:   Apollo Scientific
2,4-Hexanedione 97%
Catalog # APOSOR938383-1G
Supplier:  Apollo Scientific
CAS Number:  3002-24-2
2,4-Hexanedione 97%

Bottle, 1 g

CAS Number:  3002-24-2
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:3002-24-2
  • Formula:C₆H₁₀O₂
  • MDL Number:MFCD00009318
  • Molecular Weight:114.14 g/mol
  •  :Density: 0.9538 g/cm³ (15 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications