Order Entry
Export
Orders LinkContactUsLinkComponent
4-Ethylpyrocatechol 97%

4-Ethylpyrocatechol 97%

Bottle, 1 g
Catalog # APOSOR925237-1G
Supplier:   Apollo Scientific
4-Ethylpyrocatechol 97%
Catalog # APOSOR925237-1G
Supplier:  Apollo Scientific
CAS Number:  1124-39-6
4-Ethylpyrocatechol 97%

Bottle, 1 g

CAS Number:  1124-39-6
Price
40,60 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
  • CAS Number:1124-39-6
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00015847
  • Molecular Weight:138.17 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications