0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3-Hydroxy-2-phenyl-4(1H)-quinolinone
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%
Bottle, 5 g
APOSOR32781-5G
:
Apollo Scientific
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%
APOSOR32781-5G
:
Apollo Scientific
:
31588-18-8
3-Hydroxy-2-phenyl-4(1H)-quinolinone 95%
Bottle, 5 g
:
31588-18-8
730,00 €
1 * 5 g
-
+
:
3-Hydroxy-2-phenylquinolin-4(1H)-one
:
31588-18-8
:
C₁₅H₁₁NO₂
:
MFCD00451324
:
237.26 g/mol
:
Ambient
Pack type:
Bottle
Pk:
5 g