Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Indandione 95%

1,3-Indandione 95%

Bottle, 100 g
Catalog # APOSOR4746-100G
Irritant
Supplier:   Apollo Scientific
1,3-Indandione 95%
Catalog # APOSOR4746-100G
Supplier:  Apollo Scientific
CAS Number:  606-23-5
1,3-Indandione 95%

Bottle, 100 g

CAS Number:  606-23-5
Price
228,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,3-Indanedione, Indan-1,3-dione, 1H-Indene-1,3(2H)-dione
  • CAS Number:606-23-5
  • Formula:C₉H₆O₂
  • MDL Number:MFCD00003779
  • Molecular Weight:146.15 g/mol
  • Melting Pt:129…132 °C
  • Density:1.37 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:210-109-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications