Order Entry
Export
Orders LinkContactUsLinkComponent
1,6-Dibromohexane 97+%

1,6-Dibromohexane 97+%

Bottle, 500 g
Catalog # APOSOR4509-500G
Irritant
Health
Environment
Supplier:   Apollo Scientific
1,6-Dibromohexane 97+%
Catalog # APOSOR4509-500G
Supplier:  Apollo Scientific
CAS Number:  629-03-8
1,6-Dibromohexane 97+%

Bottle, 500 g

CAS Number:  629-03-8
Price
57,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Hexamethylen dibromide
  • CAS Number:629-03-8
  • Formula:BrH₂C(CH₂)₄CH₂Br
  • MDL Number:MFCD00000272
  • Molecular Weight:243.97 g/mol
  • Boiling Pt:242…244 °C
  • Melting Pt:–2 °C
  • Density:1.586 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:211-067-2
  • UN:3082
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications