Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,3-Dimethoxyoxetane

Bottle, 500 mg
Catalog # APOSOR312003-500MG
Irritant
Supplier:   Apollo Scientific
3,3-Dimethoxyoxetane
Catalog # APOSOR312003-500MG
Supplier:  Apollo Scientific
CAS Number:  922500-97-8
3,3-Dimethoxyoxetane

Bottle, 500 mg

CAS Number:  922500-97-8
Price
301,00 €
Log in for availability
1 * 500 mg
Product Details & Documents
  • CAS Number:922500-97-8
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD12545828
  • Molecular Weight:118.13 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg

Specifications