2,4-Dimethyloctane
Bottle, 250 mg
Catalog # APOSOR54625-250MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:4032-94-4
- Formula:C₁₀H₂₂
- MDL Number:MFCD30534470
- Molecular Weight:142.282 g/mol
- Boiling Pt:152…153 °C
- Storage Temperature:Ambient
- UN:3295
- ADR:3,III
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



