Order Entry
Export
Orders LinkContactUsLinkComponent
4-Nitroimidazole

4-Nitroimidazole

Bottle, 500 g
Catalog # APOSOR25125-500G
Irritant
Supplier:   Apollo Scientific
4-Nitroimidazole
Catalog # APOSOR25125-500G
Supplier:  Apollo Scientific
CAS Number:  3034-38-6
4-Nitroimidazole

Bottle, 500 g

CAS Number:  3034-38-6
Price
115,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole
  • CAS Number:3034-38-6
  • Formula:C₃H₃N₃O₂
  • MDL Number:MFCD00005196
  • Molecular Weight:113.08 g/mol
  • Melting Pt:303 °C (dec.)
  • Flash Pt:>200 °C
  • Storage Temperature:Ambient
  • EINECS:221-224-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications