0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
4'-Pentylacetophenone
4'-Pentylacetophenone 95%
4'-Pentylacetophenone 95%
Bottle, 5 g
APOSOR964868-5G
:
Apollo Scientific
4'-Pentylacetophenone 95%
APOSOR964868-5G
:
Apollo Scientific
:
37593-02-5
4'-Pentylacetophenone 95%
Bottle, 5 g
:
37593-02-5
33,30 €
1 * 5 g
-
+
:
p-Pentylacetophenone, 4-Pentylacetophenone
:
37593-02-5
:
C₁₃H₁₈O
:
MFCD00043687
:
190.29
:
Ambient
Pack type:
Bottle
Pk:
5 g