Order Entry
Export
Orders LinkContactUsLinkComponent
4'-Pentylacetophenone 95%

4'-Pentylacetophenone 95%

Bottle, 100 g
Catalog # APOSOR964868-100G
Supplier:   Apollo Scientific
4'-Pentylacetophenone 95%
Catalog # APOSOR964868-100G
Supplier:  Apollo Scientific
CAS Number:  37593-02-5
4'-Pentylacetophenone 95%

Bottle, 100 g

CAS Number:  37593-02-5
Price
154,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  p-Pentylacetophenone, 4-Pentylacetophenone
  • CAS Number:37593-02-5
  • Formula:C₁₃H₁₈O
  • MDL Number:MFCD00043687
  • Molecular Weight:190.29
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications