Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,4-Dimethylcyclohexanone 95%

Bottle, 1 g
Catalog # APOSOR938021-1G
Supplier:   Apollo Scientific
3,4-Dimethylcyclohexanone 95%
Catalog # APOSOR938021-1G
Supplier:  Apollo Scientific
CAS Number:  5465-09-8
3,4-Dimethylcyclohexanone 95%

Bottle, 1 g

CAS Number:  5465-09-8
Price
225,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:5465-09-8
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00045552
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications