0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
3,4-Dimethylcyclohexanone
3,4-Dimethylcyclohexanone 95%
3,4-Dimethylcyclohexanone 95%
Bottle, 10 g
APOSOR938021-10G
:
Apollo Scientific
3,4-Dimethylcyclohexanone 95%
APOSOR938021-10G
:
Apollo Scientific
:
5465-09-8
3,4-Dimethylcyclohexanone 95%
Bottle, 10 g
:
5465-09-8
992,00 €
1 * 10 g
-
+
:
5465-09-8
:
C₈H₁₄O
:
MFCD00045552
:
Ambient
Pack type:
Bottle
Pk:
10 g