Order Entry
Export
Orders LinkContactUsLinkComponent
4-(1,1-Dimethylpropyl)cyclohexanamine 96%

4-(1,1-Dimethylpropyl)cyclohexanamine 96%

Bottle, 1 g
Catalog # APOSOR951198-1G
Supplier:   Apollo Scientific
4-(1,1-Dimethylpropyl)cyclohexanamine 96%
Catalog # APOSOR951198-1G
Supplier:  Apollo Scientific
CAS Number:  208941-83-7
4-(1,1-Dimethylpropyl)cyclohexanamine 96%

Bottle, 1 g

CAS Number:  208941-83-7
Price
268,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:208941-83-7
  • Formula:C₁₁H₂₃N
  • MDL Number:MFCD03422590
  • Molecular Weight:169.31 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications