Order Entry
Export
Orders LinkContactUsLinkComponent
2-Nitrobenzaldehyde 98%

2-Nitrobenzaldehyde 98%

Bottle, 100 g
Catalog # APOSOR4976-100G
Irritant
Supplier:   Apollo Scientific
2-Nitrobenzaldehyde 98%
Catalog # APOSOR4976-100G
Supplier:  Apollo Scientific
CAS Number:  552-89-6
2-Nitrobenzaldehyde 98%

Bottle, 100 g

CAS Number:  552-89-6
Price
63,70 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:552-89-6
  • Formula:C₇H₅NO₃
  • MDL Number:MFCD00007132
  • Molecular Weight:151.12 g/mol
  • Boiling Pt:153 °C/23mmHg
  • Melting Pt:42…46 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:209-025-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications