0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
2,6-Dichloronicotinamide
2,6-Dichloronicotinamide 95%
2,6-Dichloronicotinamide 95%
2,6-Dichloronicotinamide 95%
Bottle, 5 g
APOSOR942378-5G
:
Apollo Scientific
2,6-Dichloronicotinamide 95%
APOSOR942378-5G
:
Apollo Scientific
:
62068-78-4
Expand image
2,6-Dichloronicotinamide 95%
Bottle, 5 g
Add to cart
:
62068-78-4
30,40 €
Unit of Measure
1 * 5 g
-
+
Add to cart
:
2,6-Dichloropyridine-3-carboxamide
:
62068-78-4
:
C₆H₄Cl₂N₂O
:
MFCD03230010
:
191.015 g/mol
:
Ambient
Pack type:
Bottle
Pk:
5 g