Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-(2-Hydroxyethoxy)benzaldehyde 95%

Bottle, 5 g
Catalog # APOSOR922883-5G
Irritant
Supplier:   Apollo Scientific
4-(2-Hydroxyethoxy)benzaldehyde 95%
Catalog # APOSOR922883-5G
Supplier:  Apollo Scientific
CAS Number:  22042-73-5
4-(2-Hydroxyethoxy)benzaldehyde 95%

Bottle, 5 g

CAS Number:  22042-73-5
Price
42,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:22042-73-5
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00191450
  • Molecular Weight:166.1739 g/mol
  • Melting Pt:40 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications